siRNA Details
| Gene Symbol: Slc22a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031584.2 | NP_113772.1 | 3 | 745 | ||
| GenBank mRNA | D83044.1 | - | 3 | 748 | ||
| X98334.1 | - | 3 | 694 | |||
| Y13154.1 | - | 3 | 695 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a2| Gene Symbol: Slc22a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031584.2 | NP_113772.1 | 3 | 745 | ||
| GenBank mRNA | D83044.1 | - | 3 | 748 | ||
| X98334.1 | - | 3 | 694 | |||
| Y13154.1 | - | 3 | 695 | |||