siRNA Details
| Gene Symbol: Pxmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031587.1 | NP_113775.1 | 3, 4 | 421 | ||
| GenBank mRNA | X70223.1 | - | 3, 4 | 421 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pxmp2| Gene Symbol: Pxmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031587.1 | NP_113775.1 | 3, 4 | 421 | ||
| GenBank mRNA | X70223.1 | - | 3, 4 | 421 | ||