siRNA Details
| Gene Symbol: Slc37a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031589.2 | NP_113777.2 | 3, 4 | 666 | ||
| XM_008766151.1 | XP_008764373.1 | 2, 3 | 427 | |||
| XM_008766152.1 | XP_008764374.1 | 2, 3 | 492 | |||
| XM_008766153.1 | XP_008764375.1 | 2, 3 | 427 | |||
| GenBank mRNA | AF080468.1 | - | 2, 3 | 496 | ||
| BC070889.1 | - | 3, 4 | 666 | 7098680 | ||

Slc37a4
Genomic Map