siRNA Details
| Gene Symbol: Ocln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031329.2 | NP_112619.2 | 3 | 762 | ||
| XM_006231853.2 | XP_006231915.1 | 3 | 758 | |||
| XM_006231854.2 | XP_006231916.1 | 3 | 802 | |||
| XM_006231855.2 | XP_006231917.1 | 2 | 682 | |||
| GenBank mRNA | AY033773.1 | - | 1 | 226 | ||
| BC062037.1 | - | 3 | 762 | 5623744 | ||

Ocln
Genomic Map