siRNA Details
| Gene Symbol: Slc22a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031332.1 | NP_112622.1 | 6 | 944 | ||
| XM_008760243.1 | XP_008758465.1 | 5 | 759 | |||
| GenBank mRNA | AB017446.1 | - | 6 | 944 | ||
| BC081777.1 | - | 6 | 942 | 7109603 | ||

Slc22a8
Genomic Map