siRNA Details
| Gene Symbol: Timeless | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031340.1 | NP_112630.1 | 3 | 221 | ||
| GenBank mRNA | AB019576.1 | - | 3 | 221 | ||
| BC161845.1 | - | 245 | ||||
| FQ232260.1 | - | 3 | 279 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timeless| Gene Symbol: Timeless | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031340.1 | NP_112630.1 | 3 | 221 | ||
| GenBank mRNA | AB019576.1 | - | 3 | 221 | ||
| BC161845.1 | - | 245 | ||||
| FQ232260.1 | - | 3 | 279 | |||