siRNA Details
| Gene Symbol: Slc7a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031341.1 | NP_112631.1 | 2 | 314 | ||
| XM_006252020.2 | XP_006252082.1 | 2 | 250 | |||
| XM_006252021.2 | XP_006252083.1 | 5 | 719 | |||
| XM_006252023.2 | XP_006252085.1 | 2 | 255 | |||
| XM_006252024.2 | XP_006252086.1 | 2 | 944 | |||
| XM_008770705.1 | XP_008768927.1 | 4 | 662 | |||
| GenBank mRNA | AB020520.1 | - | 2 | 314 | ||
| AF200684.1 | - | 1 | 122 | |||
| BC091142.1 | - | 2 | 229 | 7129657 | ||

Slc7a7
Genomic Map