siRNA Details
| Gene Symbol: Ppargc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031347.1 | NP_112637.1 | 8 | 976 | ||
| XM_008770219.1 | XP_008768441.1 | 8 | 1043 | |||
| XM_008770220.1 | XP_008768442.1 | 7 | 2213 | |||
| GenBank mRNA | AB025784.1 | - | 8 | 976 | ||
| AY237127.1 | - | 8 | 935 | |||

Ppargc1a
Genomic Map