siRNA Details
| Gene Symbol: Myh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031520.1 | NP_113708.1 | 2 | 514 | ||
| XM_006246821.2 | XP_006246883.1 | 2 | 561 | |||
| XM_008767880.1 | XP_008766102.1 | 2 | 671 | |||
| XM_008767881.1 | XP_008766103.1 | 1 | 357 | |||
| GenBank mRNA | AF139055.1 | - | 2 | 514 | ||

Myh10
Genomic Map