siRNA Details
| Gene Symbol: Tesk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031578.1 | NP_113766.1 | 2 | 1419 | ||
| GenBank mRNA | BC081773.1 | - | 2 | 1015 | 7131899 | |
| D50864.1 | - | 2 | 1419 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tesk1| Gene Symbol: Tesk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031578.1 | NP_113766.1 | 2 | 1419 | ||
| GenBank mRNA | BC081773.1 | - | 2 | 1015 | 7131899 | |
| D50864.1 | - | 2 | 1419 | |||