siRNA Details
| Gene Symbol: Slc37a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031589.2 | NP_113777.2 | 4, 5 | 904 | ||
| XM_008766151.1 | XP_008764373.1 | 3, 4 | 665 | |||
| XM_008766152.1 | XP_008764374.1 | 3, 4 | 730 | |||
| XM_008766153.1 | XP_008764375.1 | 3, 4 | 665 | |||
| GenBank mRNA | AF080468.1 | - | 3, 4 | 734 | ||
| BC070889.1 | - | 4, 5 | 904 | 7098680 | ||

Slc37a4
Genomic Map