siRNA Details
| Gene Symbol: Cdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031334.1 | NP_112624.1 | 13 | 2114 | ||
| GenBank mRNA | AB017696.1 | - | 13 | 2114 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh1| Gene Symbol: Cdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031334.1 | NP_112624.1 | 13 | 2114 | ||
| GenBank mRNA | AB017696.1 | - | 13 | 2114 | ||