siRNA Details
| Gene Symbol: Slc6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031343.1 | NP_112633.1 | 7, 8 | 1197 | ||
| GenBank mRNA | AB021970.1 | - | 7, 8 | 1197 | ||
| AB021971.1 | - | 7, 8 | 1197 | |||
| L29573.1 | - | 1, 2 | 138 | |||
| Y13223.1 | - | 7, 8 | 1155 | |||

Slc6a2
Genomic Map