siRNA Details
| Gene Symbol: Vdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031353.1 | NP_112643.1 | 8, 9 | 834 | ||
| XM_006246346.1 | XP_006246408.1 | 7, 8 | 807 | |||
| GenBank mRNA | AB039662.1 | - | 8, 9 | 834 | ||
| AF048828.1 | - | 8, 9 | 878 | |||
| AF268467.1 | - | 7, 8 | 768 | |||
| BC060558.1 | - | 8, 9 | 811 | |||
| BC072484.1 | - | 8, 9 | 818 | 7105146 | ||
| BC087573.1 | - | 8, 9 | 813 | 7304939 | ||
| BC087657.1 | - | 8, 9 | 790 | 7312189 | ||
| BC104684.1 | - | 8, 9 | 804 | 7462139 | ||
| BC127491.1 | - | 8, 9 | 790 | 8370617 | ||

Vdac1
Genomic Map