siRNA Details
| Gene Symbol: Ncam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031521.1 | NP_113709.1 | 7 | 1043 | ||
| XM_006242980.2 | XP_006243042.1 | 7 | 1194 | |||
| XM_006242986.2 | XP_006243048.1 | 7 | 1194 | |||
| XM_006242988.2 | XP_006243050.1 | 7 | 1194 | |||
| XM_006242994.2 | XP_006243056.1 | 7 | 1194 | |||
| XM_008766192.1 | XP_008764414.1 | 7 | 1194 | |||
| XM_008766193.1 | XP_008764415.1 | 7 | 1194 | |||
| GenBank mRNA | BC101924.1 | - | 7 | 1044 | 7936825 | |
| X06564.1 | - | 7 | 1043 | |||

Ncam1
Genomic Map