siRNA Details
| Gene Symbol: Ugt2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031533.5 | NP_113721.4 | 5 | 1273 | ||
| GenBank mRNA | BC089792.1 | - | 5 | 1256 | 7370057 | |
| DQ351215.1 | - | 5 | 1614 | |||
| X03478.1 | - | 5 | 1162 | |||
| Gene Symbol: LOC100364155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770097.1 | XP_008768319.1 | 5 | 1326 | ||
| Gene Symbol: Ugt2b17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769997.1 | XP_008768219.1 | 5 | 1613 | ||
| XM_008769998.1 | XP_008768220.1 | 5 | 2370 | |||
| XM_008769999.1 | XP_008768221.1 | 5 | 1267 | |||

Ugt2b
Genomic Map