siRNA Details
| Gene Symbol: LOC24906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031537.1 | NP_113725.1 | 6 | 601 | ||
| XM_006246376.2 | XP_006246438.1 | 6 | 651 | |||
| XM_006246377.2 | XP_006246439.1 | 6 | 681 | |||
| GenBank mRNA | AF041083.1 | - | 6 | 601 | ||

LOC24906
Genomic Map