siRNA Details
| Gene Symbol: Cyp2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031543.1 | NP_113731.1 | 5, 6 | 845 | ||
| XM_006230520.1 | XP_006230582.1 | 6, 7 | 1028 | |||
| GenBank mRNA | AF061442.1 | - | 5, 6 | 833 | ||
| BC081774.1 | - | 5, 6 | 847 | 7110096 | ||
| FQ212595.1 | - | 2, 3 | 211 | |||
| FQ218528.1 | - | 3, 4 | 484 | |||
| FQ219179.1 | - | 2, 3 | 211 | |||
| J02627.1 | - | 5, 6 | 845 | |||

Cyp2e1
Genomic Map