siRNA Details
| Gene Symbol: Nppb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031545.1 | NP_113733.1 | 2 | 258 | ||
| XM_006239376.1 | XP_006239438.1 | 2 | 308 | |||
| GenBank mRNA | FQ228997.1 | - | 314 | |||
| M25297.1 | - | 2 | 258 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nppb| Gene Symbol: Nppb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031545.1 | NP_113733.1 | 2 | 258 | ||
| XM_006239376.1 | XP_006239438.1 | 2 | 308 | |||
| GenBank mRNA | FQ228997.1 | - | 314 | |||
| M25297.1 | - | 2 | 258 | |||