siRNA Details
| Gene Symbol: Scnn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031548.2 | NP_113736.1 | 11 | 2044 | ||
| GenBank mRNA | BC070902.1 | - | 11 | 2044 | 7100777 | |
| EU627515.1 | - | 11 | 1679 | |||
| U54699.1 | - | 11 | 1711 | |||
| U54700.1 | - | 11 | 1711 | |||
| X70497.1 | - | 11 | 1757 | |||
| X70521.1 | - | 11 | 1732 | |||

Scnn1a
Genomic Map