siRNA Details
| Gene Symbol: Nfyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031553.2 | NP_113741.1 | 5 | 461 | ||
| XM_006241196.2 | XP_006241258.1 | 5 | 479 | |||
| GenBank mRNA | BC089791.1 | - | 5 | 461 | 7380395 | |
| M55045.1 | - | 4 | 330 | |||
| M60617.1 | - | 2 | 134 | |||

Nfyb
Genomic Map