siRNA Details
Gene Symbol: Ces1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031565.2 | NP_113753.2 | 13 | 1775 | ||
GenBank mRNA | X65295.1 | - | 1 | 67 | ||
X81395.1 | - | 13 | 1640 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ces1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031565.2 | NP_113753.2 | 13 | 1775 | ||
GenBank mRNA | X65295.1 | - | 1 | 67 | ||
X81395.1 | - | 13 | 1640 |