siRNA Details
| Gene Symbol: Psmc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031595.1 | NP_113783.1 | 2 | 341 | ||
| GenBank mRNA | BC062019.1 | - | 2 | 251 | 5620971 | |
| D83522.1 | - | 2 | 215 | |||
| U77918.1 | - | 2 | 341 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmc3| Gene Symbol: Psmc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031595.1 | NP_113783.1 | 2 | 341 | ||
| GenBank mRNA | BC062019.1 | - | 2 | 251 | 5620971 | |
| D83522.1 | - | 2 | 215 | |||
| U77918.1 | - | 2 | 341 | |||