siRNA Details
| Gene Symbol: Eif2ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031599.2 | NP_113787.1 | 1, 2 | 512 | ||
| XM_006236624.2 | XP_006236686.1 | 1, 2 | 550 | |||
| GenBank mRNA | AF096835.1 | - | 1, 2 | 513 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2ak3| Gene Symbol: Eif2ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031599.2 | NP_113787.1 | 1, 2 | 512 | ||
| XM_006236624.2 | XP_006236686.1 | 1, 2 | 550 | |||
| GenBank mRNA | AF096835.1 | - | 1, 2 | 513 | ||