siRNA Details
| Gene Symbol: Ywhae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031603.1 | NP_113791.1 | 2 | 305 | ||
| GenBank mRNA | U53882.1 | - | 2 | 305 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ywhae| Gene Symbol: Ywhae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031603.1 | NP_113791.1 | 2 | 305 | ||
| GenBank mRNA | U53882.1 | - | 2 | 305 | ||