siRNA Details
| Gene Symbol: Txnrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031614.2 | NP_113802.2 | 3, 4 | 332 | ||
| GenBank mRNA | AF108213.1 | - | 2, 3 | 174 | ||
| AF220760.1 | - | 2, 3 | 228 | |||
| AF220761.1 | - | 2, 3 | 206 | |||
| BC085726.1 | - | 429 | 7112649 | |||
| U63923.2 | - | 2, 3 | 223 | |||

Txnrd1
Genomic Map