siRNA Details
Gene Symbol: Grb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031623.1 | NP_113811.1 | 1, 2 | 262 | ||
GenBank mRNA | AF076619.1 | - | 1, 2 | 262 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Grb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031623.1 | NP_113811.1 | 1, 2 | 262 | ||
GenBank mRNA | AF076619.1 | - | 1, 2 | 262 |