siRNA Details
| Gene Symbol: Igbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031624.3 | NP_113812.1 | 1 | 95 | ||
| GenBank mRNA | AF000577.2 | - | 1 | 87 | ||
| BC068202.1 | - | 1 | 99 | 6921265 | ||
| FQ212697.1 | - | 1 | 104 | |||
| FQ222671.1 | - | 1 | 97 | |||

Igbp1
Genomic Map