siRNA Details
| Gene Symbol: Foxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031633.3 | NP_113821.2 | 2 | 182 | ||
| XM_006237416.2 | XP_006237478.1 | 1 | 155 | |||
| XM_006237417.2 | XP_006237479.1 | 1 | 155 | |||
| XM_006237418.2 | XP_006237480.1 | 2 | 182 | |||
| XM_006237419.2 | XP_006237481.1 | 1 | 155 | |||
| XM_006237420.2 | XP_006237482.1 | 2 | 182 | |||
| XM_006237421.2 | XP_006237483.1 | 2 | 182 | |||
| GenBank mRNA | CK469502.1 | - | 1 | 50 | ||
| U83112.1 | - | 2 | 182 | |||

Foxm1
Genomic Map