siRNA Details
Gene Symbol: Mefv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031634.1 | NP_113822.1 | 1 | 189 | ||
GenBank mRNA | AF143410.1 | - | 1 | 189 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mefv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031634.1 | NP_113822.1 | 1 | 189 | ||
GenBank mRNA | AF143410.1 | - | 1 | 189 |