siRNA Details
| Gene Symbol: Sult4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031641.1 | NP_113829.1 | 3, 4 | 429 | ||
| XM_006242114.2 | XP_006242176.1 | 3, 4 | 1221 | |||
| GenBank mRNA | AF176343.1 | - | 1 | 19 | ||
| AF188699.1 | - | 3, 4 | 429 | |||

Sult4a1
Genomic Map