siRNA Details
Gene Symbol: Hpgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031644.2 | NP_113832.1 | 3 | 534 | ||
XM_006236602.2 | XP_006236664.1 | 3 | 502 | |||
GenBank mRNA | AF021882.1 | - | 2 | 62 | ||
BC087590.1 | - | 3 | 534 | 7307804 | ||
D82071.1 | - | 3 | 365 |
