siRNA Details
| Gene Symbol: Stx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031656.2 | NP_113844.1 | 2 | 130 | ||
| XM_008767863.1 | XP_008766085.1 | 3 | 354 | |||
| XM_008767864.1 | XP_008766086.1 | 3 | 354 | |||
| XM_008767865.1 | XP_008766087.1 | 3 | 354 | |||
| GenBank mRNA | AF033109.1 | - | 2 | 135 | ||

Stx8
Genomic Map