siRNA Details
| Gene Symbol: Tgm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031659.1 | NP_113847.1 | 2, 3 | 456 | ||
| XM_006252014.2 | XP_006252076.1 | 2, 3 | 2003 | |||
| XM_008770691.1 | XP_008768913.1 | 2, 3 | 2094 | |||
| GenBank mRNA | BC097305.1 | - | 2, 3 | 412 | 7462186 | |
| FQ210559.1 | - | 2, 3 | 461 | |||
| M57263.1 | - | 2, 3 | 456 | |||

Tgm1
Genomic Map