siRNA Details
| Gene Symbol: Camkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031662.1 | NP_113850.1 | 2 | 375 | ||
| XM_006246799.2 | XP_006246861.1 | 3 | 575 | |||
| XM_006246801.2 | XP_006246863.1 | 3 | 522 | |||
| XM_008767866.1 | XP_008766088.1 | 4 | 664 | |||
| GenBank mRNA | AB023658.1 | - | 2 | 375 | ||
| L42810.1 | - | 2 | 375 | |||
| S83194.1 | - | 2 | 375 | |||

Camkk1
Genomic Map