siRNA Details
| Gene Symbol: Smc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031683.1 | NP_113871.1 | 3, 4 | 470 | ||
| GenBank mRNA | AJ005113.1 | - | 3, 4 | 470 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smc1a| Gene Symbol: Smc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031683.1 | NP_113871.1 | 3, 4 | 470 | ||
| GenBank mRNA | AJ005113.1 | - | 3, 4 | 470 | ||