siRNA Details
| Gene Symbol: Ykt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031692.2 | NP_113880.2 | 1, 2 | 106 | ||
| GenBank mRNA | AF033027.1 | - | 1, 2 | 106 | ||
| AY881621.1 | - | 1, 2 | 106 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ykt6| Gene Symbol: Ykt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031692.2 | NP_113880.2 | 1, 2 | 106 | ||
| GenBank mRNA | AF033027.1 | - | 1, 2 | 106 | ||
| AY881621.1 | - | 1, 2 | 106 | |||