siRNA Details
| Gene Symbol: Hsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031694.2 | NP_113882.1 | 3 | 434 | ||
| XM_006256505.2 | XP_006256567.1 | 3 | 354 | |||
| XM_006256506.2 | XP_006256568.1 | 3 | 589 | |||
| GenBank mRNA | AF172640.1 | - | 3 | 286 | ||
| BC086554.1 | - | 3 | 434 | 7302950 | ||

Hsf2
Genomic Map