siRNA Details
| Gene Symbol: Psmc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031595.1 | NP_113783.1 | 4 | 560 | ||
| GenBank mRNA | BC062019.1 | - | 4 | 470 | 5620971 | |
| D83522.1 | - | 4 | 434 | |||
| FQ225567.1 | - | 3 | 219 | |||
| FQ225947.1 | - | 3 | 219 | |||
| FQ226273.1 | - | 3 | 219 | |||
| FQ232987.1 | - | 3 | 219 | |||
| FQ233836.1 | - | 3 | 219 | |||
| U77918.1 | - | 4 | 560 | |||

Psmc3
Genomic Map