siRNA Details
| Gene Symbol: Eif2ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031599.2 | NP_113787.1 | 2, 3 | 642 | ||
| XM_006236623.1 | XP_006236685.1 | 2, 3 | 160 | |||
| XM_006236624.2 | XP_006236686.1 | 2, 3 | 680 | |||
| XM_008762977.1 | XP_008761199.1 | 1, 2 | 274 | |||
| GenBank mRNA | AF096835.1 | - | 2, 3 | 643 | ||

Eif2ak3
Genomic Map