siRNA Details
| Gene Symbol: Igbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031624.3 | NP_113812.1 | 1 | 176 | ||
| GenBank mRNA | AF000577.2 | - | 1 | 168 | ||
| BC068202.1 | - | 1 | 180 | 6921265 | ||
| FQ212697.1 | - | 1 | 185 | |||
| FQ222671.1 | - | 1 | 178 | |||

Igbp1
Genomic Map