siRNA Details
Gene Symbol: Psmb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031629.1 | NP_113817.1 | 3 | 479 | ||
GenBank mRNA | L17127.1 | - | 3 | 479 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Psmb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031629.1 | NP_113817.1 | 3 | 479 | ||
GenBank mRNA | L17127.1 | - | 3 | 479 |