siRNA Details
| Gene Symbol: Vapa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031631.2 | NP_113819.3 | 4 | 584 | ||
| XM_008767384.1 | XP_008765606.1 | 4 | 1137 | |||
| GenBank mRNA | AF086630.1 | - | 4 | 332 | ||
| BC061875.1 | - | 4 | 584 | 5598274 | ||
| FQ228815.1 | - | 4 | 441 | |||
| FQ234023.1 | - | 4 | 341 | |||

Vapa
Genomic Map