siRNA Details
| Gene Symbol: Cpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031640.1 | NP_113828.1 | 3, 4 | 561 | ||
| XM_008765417.1 | XP_008763639.1 | 2, 3 | 662 | |||
| GenBank mRNA | AF097723.1 | - | 3, 4 | 561 | ||
| AF131077.1 | - | 2, 3 | 503 | |||
| BC070884.1 | - | 2, 3 | 522 | 7098277 | ||
| FQ210652.1 | - | 1, 2 | 301 | |||

Cpq
Genomic Map