siRNA Details
| Gene Symbol: Sult4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031641.1 | NP_113829.1 | 4 | 456 | ||
| XM_006242114.2 | XP_006242176.1 | 4 | 1248 | |||
| GenBank mRNA | AF176343.1 | - | 1 | 46 | ||
| AF188699.1 | - | 4 | 456 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult4a1| Gene Symbol: Sult4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031641.1 | NP_113829.1 | 4 | 456 | ||
| XM_006242114.2 | XP_006242176.1 | 4 | 1248 | |||
| GenBank mRNA | AF176343.1 | - | 1 | 46 | ||
| AF188699.1 | - | 4 | 456 | |||