siRNA Details
Gene Symbol: Sfmbt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031647.2 | NP_113835.2 | 4 | 403 | ||
XM_006252656.2 | XP_006252718.1 | 4 | 689 | |||
XM_006252657.2 | XP_006252719.1 | 5 | 749 | |||
XM_006252658.1 | XP_006252720.1 | 4 | 2661 | |||
XM_008771003.1 | XP_008769225.1 | 6 | 1670 | |||
XM_008771004.1 | XP_008769226.1 | 5 | 1610 | |||
XM_008771005.1 | XP_008769227.1 | 7 | 979 | |||
XM_008771006.1 | XP_008769228.1 | 6 | 843 | |||
GenBank mRNA | AB032164.1 | - | 4 | 397 | ||
BC081770.1 | - | 4 | 403 | 7132495 |
