siRNA Details
| Gene Symbol: Ncan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031653.2 | NP_113841.2 | 3 | 348 | ||
| GenBank mRNA | M97161.1 | - | 3 | 348 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncan| Gene Symbol: Ncan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031653.2 | NP_113841.2 | 3 | 348 | ||
| GenBank mRNA | M97161.1 | - | 3 | 348 | ||