siRNA Details
| Gene Symbol: Mybbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031668.1 | NP_113856.1 | 3, 4 | 254 | ||
| XM_006246802.1 | XP_006246864.1 | 4, 5 | 498 | |||
| GenBank mRNA | U83590.1 | - | 3, 4 | 254 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mybbp1a| Gene Symbol: Mybbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031668.1 | NP_113856.1 | 3, 4 | 254 | ||
| XM_006246802.1 | XP_006246864.1 | 4, 5 | 498 | |||
| GenBank mRNA | U83590.1 | - | 3, 4 | 254 | ||