siRNA Details
Gene Symbol: P2ry4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031680.1 | NP_113868.1 | 2 | 626 | ||
XM_006257093.2 | XP_006257155.1 | 4 | 1266 | |||
XM_006257094.2 | XP_006257156.1 | 3 | 835 | |||
XM_006257095.2 | XP_006257157.1 | 3 | 1151 | |||
XM_008773269.1 | XP_008771491.1 | 5 | 984 | |||
XM_008773270.1 | XP_008771492.1 | 5 | 966 | |||
GenBank mRNA | Y11433.1 | - | 2 | 626 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
