siRNA Details
| Gene Symbol: Slc29a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031684.2 | NP_113872.1 | 11 | 1107 | ||
| XM_006244560.1 | XP_006244622.1 | 11 | 1138 | |||
| XM_006244561.2 | XP_006244623.1 | 12 | 1371 | |||
| XM_006244562.2 | XP_006244624.1 | 11 | 1310 | |||
| XM_006244563.2 | XP_006244625.1 | 11 | 1133 | |||
| XM_008766879.1 | XP_008765101.1 | 12 | 1196 | |||
| GenBank mRNA | AF015304.1 | - | 8 | 793 | ||
| BC078789.1 | - | 11 | 1107 | 7132212 | ||

Slc29a1
Genomic Map